Product Details
- SNP ID
-
rs200683677
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:135287702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGATGCCATTTCTGTACACTGTGT[G/T]TATCACCAGGATTTTCCCTGCCCAT
- Phenotype
-
MIM: 314997
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF75D
PubMed Links
Gene Details
- Gene
- ZNF75D
- Gene Name
- zinc finger protein 75D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001185063.2 |
2807 |
Intron |
|
|
NP_001171992.1 |
NM_007131.4 |
2807 |
Missense Mutation |
AAA,ACA |
K323T |
NP_009062.2 |
XM_005262469.1 |
2807 |
Missense Mutation |
AAA,ACA |
K323T |
XP_005262526.1 |
XM_005262471.3 |
2807 |
Missense Mutation |
AAA,ACA |
K116T |
XP_005262528.1 |
XM_011531391.1 |
2807 |
Missense Mutation |
AAA,ACA |
K323T |
XP_011529693.1 |
XM_011531395.2 |
2807 |
Missense Mutation |
AAA,ACA |
K173T |
XP_011529697.1 |
XM_011531396.2 |
2807 |
Missense Mutation |
AAA,ACA |
K155T |
XP_011529698.1 |
XM_011531397.2 |
2807 |
Missense Mutation |
AAA,ACA |
K152T |
XP_011529699.1 |
XM_017029818.1 |
2807 |
Missense Mutation |
AAA,ACA |
K177T |
XP_016885307.1 |
XM_017029819.1 |
2807 |
Missense Mutation |
AAA,ACA |
K189T |
XP_016885308.1 |
XM_017029820.1 |
2807 |
Missense Mutation |
AAA,ACA |
K152T |
XP_016885309.1 |
View Full Product Details