Product Details

SNP ID
rs201126736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130384497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGTGACAGGAACTTTCCCTGGG[A/C]CTCTCTGGGCCACAATTCCTGGCCG
Phenotype
MIM: 300513
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR119 PubMed Links

Gene Details

Gene
GPR119
Gene Name
G protein-coupled receptor 119
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178471.2 1041 Missense Mutation AGG,AGT R317S NP_848566.1

View Full Product Details