Product Details

SNP ID
rs1477941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:11692441 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGCGGTAGCAGAGCTCTCCCCT[C/G]CAGTGCTCACCTCCAGAATATCGCA
Phenotype
MIM: 139312
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNAL PubMed Links

Gene Details

Gene
GNAL
Gene Name
G protein subunit alpha L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142339.2 Intron NP_001135811.1
NM_001261443.1 Intron NP_001248372.1
NM_001261444.1 Intron NP_001248373.1
NM_182978.3 Intron NP_892023.1
XM_006722324.2 Intron XP_006722387.1

View Full Product Details