Product Details

SNP ID
rs35925424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:58203637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGATTTTGAGATAGAAAATCAT[A/C]ACGGAGCATCCAGGTTGATTAAATA
Phenotype
MIM: 616070 MIM: 115442
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC113 PubMed Links

Gene Details

Gene
CCDC113
Gene Name
coiled-coil domain containing 113
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142302.1 Intron NP_001135774.1
NM_014157.3 Intron NP_054876.2
XM_011523038.2 Intron XP_011521340.1
XM_011523039.2 Intron XP_011521341.1
XM_011523040.2 Intron XP_011521342.1
XM_011523041.2 Intron XP_011521343.1
XM_011523042.2 Intron XP_011521344.1
XM_017023176.1 Intron XP_016878665.1
XM_017023177.1 Intron XP_016878666.1
XM_017023178.1 Intron XP_016878667.1
Gene
CSNK2A2
Gene Name
casein kinase 2 alpha 2
There are no transcripts associated with this gene.

View Full Product Details