Product Details

SNP ID
rs4629709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:129833541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCTTCCACAACTGGAACAAAAA[A/G]CAATCCACAAACAAAGTAGGTGACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM244 PubMed Links

Gene Details

Gene
TMEM244
Gene Name
transmembrane protein 244
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010876.1 494 Missense Mutation CTT,TTT L80F NP_001010876.1
XM_011535669.2 494 Missense Mutation CTT,TTT L80F XP_011533971.1

View Full Product Details