Product Details
- SNP ID
-
rs4624895
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:129830388 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GTAGAATCTTTGATGCACCCGTGGG[A/C]ATATAATCAAGTGTGTATGTAGATT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM244
PubMed Links
Gene Details
- Gene
- TMEM244
- Gene Name
- transmembrane protein 244
View Full Product Details