Product Details

SNP ID
rs4690097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:3444018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCCTCCCAGTCCGCCCCTCACA[C/T]CCCCTCCCGCATGTCCCCAGCTGCC
Phenotype
MIM: 604552 MIM: 602512
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HGFAC PubMed Links

Gene Details

Gene
HGFAC
Gene Name
HGF activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297439.1 Intron NP_001284368.1
NM_001528.3 Intron NP_001519.1
Gene
RGS12
Gene Name
regulator of G-protein signaling 12
There are no transcripts associated with this gene.

View Full Product Details