Product Details

SNP ID
rs2272484
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:52087533 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTTGCTCCAGCGGCTGCTCTCCTGG[C/T]CACATCTGCTGTGGCTGACCCCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7E47P PubMed Links

Gene Details

Gene
OR7E47P
Gene Name
olfactory receptor family 7 subfamily E member 47 pseudogene
There are no transcripts associated with this gene.

View Full Product Details