Product Details

SNP ID
rs3802936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65128765 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTGGCCTTGGCATCCAAGGTTG[A/G]GCAGATTCTCCTGCCCTTGGTGAGC
Phenotype
MIM: 134690 MIM: 606866 MIM: 608046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAU PubMed Links

Gene Details

Gene
FAU
Gene Name
FAU, ubiquitin like and ribosomal protein S30 fusion
There are no transcripts associated with this gene.

Gene
MIR6751
Gene Name
microRNA 6751
There are no transcripts associated with this gene.

Gene
MRPL49
Gene Name
mitochondrial ribosomal protein L49
There are no transcripts associated with this gene.

Gene
SYVN1
Gene Name
synoviolin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032431.2 Intron NP_115807.1
NM_172230.2 Intron NP_757385.1
XM_011545303.2 Intron XP_011543605.1

View Full Product Details