Product Details

SNP ID
rs1319915
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:97776258 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAATTCTCCGGGTGCCGCGTCCGCA[C/T]TTCCCCCGGCTGGGCCAGCGCCGAC
Phenotype
MIM: 613296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAPTM4B PubMed Links

Gene Details

Gene
LAPTM4B
Gene Name
lysosomal protein transmembrane 4 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018407.4 Intron NP_060877.3

View Full Product Details