Product Details
- SNP ID
-
rs382367
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52469837 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCTTGTTCTTAGCATCCTTGAG[T/G]GCCATCTCCCCACGCTGCTCAGCAT
- Phenotype
-
MIM: 612315
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT6C
PubMed Links
Gene Details
- Gene
- KRT6C
- Gene Name
- keratin 6C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173086.4 |
1305 |
Silent Mutation |
GCA,GCC |
A419A |
NP_775109.2 |
View Full Product Details