Product Details

SNP ID
rs401641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39606399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTTGACCTCGGCCAACGTGGC[C/G]TCCGTCCCCTCTTCTCCACGGAGAG
Phenotype
MIM: 601725
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NEUROD2 PubMed Links

Gene Details

Gene
NEUROD2
Gene Name
neuronal differentiation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006160.3 401 Missense Mutation GAC,GAG D67E NP_006151.3

View Full Product Details