Product Details

SNP ID
rs17622614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123686281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTGGAAAGGAGTTCAATTGAG[C/T]CATGTTAATTTCTGTTTACTCTTCT
Phenotype
MIM: 605056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WASL PubMed Links

Gene Details

Gene
WASL
Gene Name
Wiskott-Aldrich syndrome like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003941.3 Intron NP_003932.3

View Full Product Details