Product Details

SNP ID
rs11250129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11344645 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATTGTTGGGCAGGAAAACCTCC[A/G]GGGTGCTGCGCTCTTCCCAGCGCCT
Phenotype
MIM: 615174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TDH PubMed Links

Gene Details

Gene
TDH
Gene Name
L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details