Product Details

SNP ID
rs12709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200853385 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTAATTCTTTTGGCTGGATCATAC[C/T]CCAACATTTTCTGAATGAGGTCAAA
Phenotype
MIM: 601951
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLK1 PubMed Links

Gene Details

Gene
CLK1
Gene Name
CDC like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162407.1 1557 Missense Mutation GAG,GGG E501G NP_001155879.1
NM_004071.3 1557 Missense Mutation GAG,GGG E459G NP_004062.2

View Full Product Details