Product Details

SNP ID
rs2042295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53795721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCACACAGCCAGTAGATGACATAG[C/G]TAATATGTGGCCTCATCTGTATGCC
Phenotype
MIM: 612043 MIM: 612044 MIM: 611954 MIM: 609648
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR371A PubMed Links

Gene Details

Gene
MIR371A
Gene Name
microRNA 371a
There are no transcripts associated with this gene.

Gene
MIR371B
Gene Name
microRNA 371b
There are no transcripts associated with this gene.

Gene
MIR372
Gene Name
microRNA 372
There are no transcripts associated with this gene.

Gene
MIR373
Gene Name
microRNA 373
There are no transcripts associated with this gene.

Gene
NLRP12
Gene Name
NLR family pyrin domain containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277126.1 Intron NP_001264055.1
NM_001277129.1 Intron NP_001264058.1
NM_144687.3 Intron NP_653288.1
XM_011527479.1 Intron XP_011525781.1
XM_011527480.1 Intron XP_011525782.1
XM_011527482.1 Intron XP_011525784.1
XM_017027460.1 Intron XP_016882949.1
XM_017027461.1 Intron XP_016882950.1
XM_017027462.1 Intron XP_016882951.1
XM_017027463.1 Intron XP_016882952.1
XM_017027464.1 Intron XP_016882953.1
XM_017027465.1 Intron XP_016882954.1
XM_017027466.1 Intron XP_016882955.1
XM_017027467.1 Intron XP_016882956.1

View Full Product Details