Product Details

SNP ID
rs2982299
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:25623284 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CAACTTAACTCCCACACGTTGGTTG[G/T]GTCTCTGCTTTCATTTCTAGTTGTA
Phenotype
MIM: 616772
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAN1C1 PubMed Links

Gene Details

Gene
MAN1C1
Gene Name
mannosidase alpha class 1C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289010.1 Intron NP_001275939.1
NM_020379.3 Intron NP_065112.1
XM_017001860.1 Intron XP_016857349.1
XM_017001861.1 Intron XP_016857350.1

View Full Product Details