Product Details

SNP ID
rs7333332
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:95580429 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATATAGTGACTTAATTTAAATACTG[C/G]GTTTACTTAGCAAAAGTTCATTTCC
Phenotype
MIM: 608671
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 5264 Intron NP_001153572.1
NM_006984.4 5264 Intron NP_008915.1
NM_182848.3 5264 Intron NP_878268.1
XM_011521134.2 5264 Intron XP_011519436.1
XM_017020843.1 5264 Intron XP_016876332.1
XM_017020844.1 5264 Intron XP_016876333.1
Gene
DZIP1
Gene Name
DAZ interacting zinc finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014934.4 5264 UTR 3 NP_055749.1
NM_198968.3 5264 UTR 3 NP_945319.1
XM_005254032.1 5264 Intron XP_005254089.1
XM_006719916.1 5264 Intron XP_006719979.1
XM_006719917.2 5264 Intron XP_006719980.1
XM_006719918.1 5264 Intron XP_006719981.1
XM_006719920.1 5264 Intron XP_006719983.1
XM_017020444.1 5264 Intron XP_016875933.1
XM_017020445.1 5264 Intron XP_016875934.1
XM_017020446.1 5264 Intron XP_016875935.1
XM_017020447.1 5264 Intron XP_016875936.1

View Full Product Details