Product Details

SNP ID
rs1886888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53227398 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAAAGATTAAACAACTTGCCCA[A/G]TGATACACAGGAAGTTGCTGACTCT
Phenotype
MIM: 300522 MIM: 314690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQSEC2 PubMed Links

Gene Details

Gene
IQSEC2
Gene Name
IQ motif and Sec7 domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111125.2 4292 Intron NP_001104595.1
NM_001243197.1 4292 Intron NP_001230126.1
NM_015075.1 4292 Intron NP_055890.1
XM_006724579.2 4292 Intron XP_006724642.1
XM_006724580.3 4292 Intron XP_006724643.1
XM_006724581.3 4292 Intron XP_006724644.1
XM_006724582.3 4292 UTR 3 XP_006724645.1
XM_006724583.3 4292 Intron XP_006724646.1
XM_006724584.2 4292 Intron XP_006724647.1
XM_011530773.2 4292 Intron XP_011529075.1
XM_011530774.2 4292 Intron XP_011529076.1
XM_011530776.1 4292 Intron XP_011529078.1
XM_011530777.1 4292 Intron XP_011529079.1
XM_017029359.1 4292 Intron XP_016884848.1
XM_017029360.1 4292 Intron XP_016884849.1
Gene
KDM5C
Gene Name
lysine demethylase 5C
There are no transcripts associated with this gene.

View Full Product Details