Product Details

SNP ID
rs2057464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:108026521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTAGGAAGCTGTGTTGTTTCCA[C/T]AGCATGCTAAAATCATAAACCAAAC
Phenotype
MIM: 300620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VSIG1 PubMed Links

Gene Details

Gene
VSIG1
Gene Name
V-set and immunoglobulin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170553.1 Intron NP_001164024.1
NM_182607.4 Intron NP_872413.1
XM_005262127.4 Intron XP_005262184.1
XM_011530936.2 Intron XP_011529238.1

View Full Product Details