Product Details

SNP ID
rs823139
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:205770268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATTCAAATGTACTTAATAAAAT[G/T]GTCAGGTGGGCAACCAAAGGGAAGA
Phenotype
MIM: 603949
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB29 PubMed Links

Gene Details

Gene
RAB29
Gene Name
RAB29, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135662.1 632 UTR 3 NP_001129134.1
NM_001135663.1 632 UTR 3 NP_001129135.1
NM_001135664.1 632 UTR 3 NP_001129136.1
NM_003929.2 632 UTR 3 NP_003920.1
XM_005245569.1 632 UTR 3 XP_005245626.1
XM_005245570.1 632 UTR 3 XP_005245627.1
XM_005245571.1 632 UTR 3 XP_005245628.1
XM_006711605.3 632 UTR 3 XP_006711668.1
XM_006711606.2 632 UTR 3 XP_006711669.1
XM_017002748.1 632 UTR 3 XP_016858237.1
XM_017002749.1 632 UTR 3 XP_016858238.1
XM_017002750.1 632 UTR 3 XP_016858239.1

View Full Product Details