Product Details
- SNP ID
-
rs7762170
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:122994952 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGTCATAATCACTTAAAATCCTA[A/G]AAGGACCTTATATATAGTGCATATA
- Phenotype
-
MIM: 616945
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLVS2
PubMed Links
Gene Details
- Gene
- CLVS2
- Gene Name
- clavesin 2
View Full Product Details