Product Details

SNP ID
rs2288906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38212137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGGTGACAGCTCCAGCTCCCTG[C/T]GGGGGCAGCCGAAGCTGAAGTCAGG
Phenotype
MIM: 601670 MIM: 616655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPF1 PubMed Links

Gene Details

Gene
DPF1
Gene Name
double PHD fingers 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135155.2 Intron NP_001128627.1
NM_001135156.2 Intron NP_001128628.1
NM_001289978.1 Intron NP_001276907.1
NM_004647.3 Intron NP_004638.2
XM_005259289.2 Intron XP_005259346.1
XM_005259290.3 Intron XP_005259347.1
XM_005259292.4 Intron XP_005259349.1
XM_006723407.2 Intron XP_006723470.1
XM_006723408.3 Intron XP_006723471.1
XM_006723409.2 Intron XP_006723472.1
XM_006723410.2 Intron XP_006723473.1
XM_011527356.1 Intron XP_011525658.1
XM_011527357.2 Intron XP_011525659.1
XM_011527358.2 Intron XP_011525660.1
Gene
SIPA1L3
Gene Name
signal induced proliferation associated 1 like 3
There are no transcripts associated with this gene.

View Full Product Details