Product Details

SNP ID
rs9565497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:79406330 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCGGAGAATGCGCGGGTCGAGTG[C/G]TGTAGTTCGATTTTTGCGTGTGTGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBM26 PubMed Links

Gene Details

Gene
RBM26
Gene Name
RNA binding motif protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286631.1 Intron NP_001273560.1
NM_001286632.1 Intron NP_001273561.1
NM_022118.4 Intron NP_071401.3
XM_005266491.4 Intron XP_005266548.1
XM_005266497.1 Intron XP_005266554.1
XM_006719857.1 Intron XP_006719920.1
XM_011535179.1 Intron XP_011533481.1
XM_011535180.2 Intron XP_011533482.1
XM_011535181.1 Intron XP_011533483.1
XM_011535182.1 Intron XP_011533484.1
XM_011535183.2 Intron XP_011533485.1
XM_011535184.1 Intron XP_011533486.1
XM_011535185.2 Intron XP_011533487.1
XM_011535186.2 Intron XP_011533488.1
XM_011535187.2 Intron XP_011533489.1
XM_011535188.2 Intron XP_011533490.1
XM_011535189.1 Intron XP_011533491.1
XM_011535190.1 Intron XP_011533492.1
XM_011535191.2 Intron XP_011533493.1
XM_011535192.2 Intron XP_011533494.1
XM_011535193.2 Intron XP_011533495.1
XM_011535195.1 Intron XP_011533497.1
XM_017020688.1 Intron XP_016876177.1
XM_017020689.1 Intron XP_016876178.1
XM_017020690.1 Intron XP_016876179.1
XM_017020691.1 Intron XP_016876180.1
XM_017020692.1 Intron XP_016876181.1
XM_017020693.1 Intron XP_016876182.1
XM_017020694.1 Intron XP_016876183.1
XM_017020695.1 Intron XP_016876184.1
XM_017020696.1 Intron XP_016876185.1
XM_017020697.1 Intron XP_016876186.1
XM_017020698.1 Intron XP_016876187.1
XM_017020699.1 Intron XP_016876188.1
XM_017020700.1 Intron XP_016876189.1
XM_017020701.1 Intron XP_016876190.1
XM_017020702.1 Intron XP_016876191.1
XM_017020703.1 Intron XP_016876192.1
Gene
RBM26-AS1
Gene Name
RBM26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details