Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000144.4 | Intron | NP_000135.2 | |||
NM_001161706.1 | Intron | NP_001155178.1 | |||
NM_181425.2 | Intron | NP_852090.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170414.2 | Intron | NP_001163885.1 | |||
NM_001170415.1 | Intron | NP_001163886.1 | |||
NM_001170416.1 | Intron | NP_001163887.1 | |||
NM_004817.3 | Intron | NP_004808.2 | |||
NM_201629.3 | Intron | NP_963923.1 | |||
XM_005252314.2 | Intron | XP_005252371.1 | |||
XM_011519204.1 | Intron | XP_011517506.1 | |||
XM_011519206.2 | Intron | XP_011517508.1 | |||
XM_011519207.2 | Intron | XP_011517509.1 | |||
XM_011519208.2 | Intron | XP_011517510.1 | |||
XM_011519209.2 | Intron | XP_011517511.1 | |||
XM_017015326.1 | Intron | XP_016870815.1 | |||
XM_017015327.1 | Intron | XP_016870816.1 | |||
XM_017015328.1 | Intron | XP_016870817.1 |