Product Details

SNP ID
rs12671608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:153827030 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCTTTATGACATTTTCTGGGAAA[C/T]TTATGACTACCAAGTGTTGAAGACC
Phenotype
MIM: 126141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPP6 PubMed Links

Gene Details

Gene
DPP6
Gene Name
dipeptidyl peptidase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039350.2 Intron NP_001034439.1
NM_001290252.1 Intron NP_001277181.1
NM_001290253.1 Intron NP_001277182.1
NM_001936.4 Intron NP_001927.3
NM_130797.3 Intron NP_570629.2
XM_017011808.1 Intron XP_016867297.1
XM_017011809.1 Intron XP_016867298.1
XM_017011810.1 Intron XP_016867299.1
XM_017011811.1 Intron XP_016867300.1
XM_017011812.1 Intron XP_016867301.1

View Full Product Details