Product Details

SNP ID
rs10521485
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:56993429 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTGCAGATACCAGCCAGAGCTATT[A/G]GAGACAAGAGAGACCCTAATGCCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPIN3 PubMed Links

Gene Details

Gene
SPIN3
Gene Name
spindlin family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010862.2 1829 UTR 3 NP_001010862.2
XM_006724577.3 1829 UTR 3 XP_006724640.1
XM_006724578.3 1829 UTR 3 XP_006724641.1
XM_017029315.1 1829 UTR 3 XP_016884804.1

View Full Product Details