Product Details

SNP ID
rs676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:16238735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACCTCAAGCTCGACTTCAAGGA[C/T]GTCCTGCTCCGACCTAAGCGGAGCA
Phenotype
MIM: 139265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMPR PubMed Links

Gene Details

Gene
GMPR
Gene Name
guanosine monophosphate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006877.3 144 Silent Mutation GAC,GAT D14D NP_006868.3
XM_011514508.2 144 Silent Mutation GAC,GAT D14D XP_011512810.1

View Full Product Details