Product Details

SNP ID
rs11488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10133368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGTTTATAGGAGAGATTTATT[A/T]GAAGAAATATTACAACATATAAAAA
Phenotype
MIM: 126375
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNMT1 PubMed Links

Gene Details

Gene
DNMT1
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130823.2 5434 UTR 3 NP_001124295.1
NM_001318730.1 5434 UTR 3 NP_001305659.1
NM_001318731.1 5434 UTR 3 NP_001305660.1
NM_001379.3 5434 UTR 3 NP_001370.1

View Full Product Details