Product Details

SNP ID
rs2033971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158545334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCACAACGGCACAGAATCATCA[A/C]GTGCCTGGGACTGGCCTCCCCTGGC
Phenotype
MIM: 613209
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM181 PubMed Links

Gene Details

Gene
TMEM181
Gene Name
transmembrane protein 181
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020823.1 Intron NP_065874.1
XM_005267074.3 Intron XP_005267131.1
XM_011535999.1 Intron XP_011534301.1
XM_011536000.1 Intron XP_011534302.1
XM_011536001.2 Intron XP_011534303.1
XM_011536002.2 Intron XP_011534304.1
XM_011536003.2 Intron XP_011534305.1

View Full Product Details