Product Details

SNP ID
rs3207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144930488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAATCTTCTCCCTTGTAACAAA[C/T]GTGCAGTCCGTTCACAAGCTGTAAA
Phenotype
MIM: 601262
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF16 PubMed Links

Gene Details

Gene
ZNF16
Gene Name
zinc finger protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029976.2 2485 UTR 3 NP_001025147.2
NM_006958.2 2485 UTR 3 NP_008889.2
XM_005272341.2 2485 UTR 3 XP_005272398.1
XM_005272342.2 2485 UTR 3 XP_005272399.1
XM_011517298.1 2485 UTR 3 XP_011515600.1

View Full Product Details