Product Details

SNP ID
rs2054515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57551395 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCATGGATCTGAACCTAAATCTGT[A/G]ACTGCAAACCTCCCATCTCTTCTAT
Phenotype
MIM: 607376 MIM: 602821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN2 PubMed Links

Gene Details

Gene
DCTN2
Gene Name
dynactin subunit 2
There are no transcripts associated with this gene.

Gene
KIF5A
Gene Name
kinesin family member 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004984.2 Intron NP_004975.2

View Full Product Details