Product Details

SNP ID
rs1920606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52088244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAATGGAAAGACAGCAGAGTGAC[A/G]GGATGGAAACCTGTGGGAGACTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7E47P PubMed Links

Gene Details

Gene
OR7E47P
Gene Name
olfactory receptor family 7 subfamily E member 47 pseudogene
There are no transcripts associated with this gene.

View Full Product Details