Product Details

SNP ID
rs2803849
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:235339932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTATCTATCATTCCTTTGTAT[A/G]TGACTACAATGACTTCTAGTGGTAA
Phenotype
MIM: 606982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GGPS1 PubMed Links

Gene Details

Gene
GGPS1
Gene Name
geranylgeranyl diphosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037277.1 Intron NP_001032354.1

View Full Product Details