Product Details

SNP ID
rs2449272
Assay Type
Validated
NCBI dbSNP Submissions
34
Location
Chr.1:224929311 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACGCCCACGAGTTGTGAAAATCTTA[C/A]GTGTAAGTCACCTCTTTCAGAGGTC
Phenotype
MIM: 603341
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145154.1 889 Intron NP_001138626.1
NM_001373.1 889 Intron NP_001364.1
NM_144989.2 889 Intron NP_659426.2
XM_006711735.3 889 UTR 5 XP_006711798.1
XM_011544057.2 889 UTR 5 XP_011542359.1
XM_011544058.2 889 UTR 5 XP_011542360.1
XM_011544059.2 889 UTR 5 XP_011542361.1
XM_011544061.2 889 UTR 5 XP_011542363.1
XM_011544062.2 889 UTR 5 XP_011542364.1
XM_011544063.2 889 UTR 5 XP_011542365.1
XM_011544064.2 889 UTR 5 XP_011542366.1
XM_011544065.2 889 UTR 5 XP_011542367.1
XM_011544066.2 889 UTR 5 XP_011542368.1
XM_011544067.2 889 UTR 5 XP_011542369.1
XM_011544069.2 889 UTR 5 XP_011542371.1
XM_011544070.2 889 UTR 5 XP_011542372.1
XM_011544071.2 889 UTR 5 XP_011542373.1
XM_011544072.2 889 UTR 5 XP_011542374.1
XM_011544073.2 889 Intron XP_011542375.1
XM_011544074.2 889 Intron XP_011542376.1
XM_011544076.1 889 Intron XP_011542378.1
XM_011544079.1 889 Intron XP_011542381.1
XM_011544081.1 889 Intron XP_011542383.1
XM_017000295.1 889 UTR 5 XP_016855784.1
XM_017000296.1 889 UTR 5 XP_016855785.1
XM_017000297.1 889 UTR 5 XP_016855786.1
XM_017000298.1 889 UTR 5 XP_016855787.1
XM_017000299.1 889 Intron XP_016855788.1
XM_017000300.1 889 UTR 5 XP_016855789.1

View Full Product Details