Product Details
- SNP ID
-
hCV15821407
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44799410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGGATTTGGTTATAAGGGTTCCT[A/G]CTTTCACAGAATTATTCCAGGGTTT
- Phenotype
-
MIM: 123840
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PPIA
PubMed Links
Gene Details
- Gene
- PPIA
- Gene Name
- peptidylprolyl isomerase A
View Full Product Details