Product Details

SNP ID
rs2831139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:27727935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTACATTAGCTGTTGTTATATT[G/T]TCAGATTTAATAAAATAATCAAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00113 PubMed Links

Gene Details

Gene
LINC00113
Gene Name
long intergenic non-protein coding RNA 113
There are no transcripts associated with this gene.

View Full Product Details