Product Details

SNP ID
rs2148275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89706437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCAATTTAACATTTTCTTTTCTT[C/T]TTTGGTGAATTTTTTTGTGTGTGTG
Phenotype
MIM: 605498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ37201 PubMed Links

Gene Details

Gene
FLJ37201
Gene Name
tigger transposable element derived 2 pseudogene
There are no transcripts associated with this gene.

Gene
KIF20B
Gene Name
kinesin family member 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284259.1 Intron NP_001271188.1
NM_016195.3 Intron NP_057279.2

View Full Product Details