Product Details

SNP ID
rs2888131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:213001990 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAATTAAAGTAAGTAAAATCTTG[A/G]TAAGTGCATAGTAGGGGATTCAGTG
Phenotype
MIM: 606234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IKZF2 PubMed Links

Gene Details

Gene
IKZF2
Gene Name
IKAROS family zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079526.1 7163 UTR 3 NP_001072994.1
NM_016260.2 7163 UTR 3 NP_057344.2
XM_005246384.4 7163 UTR 3 XP_005246441.1
XM_005246385.3 7163 UTR 3 XP_005246442.1
XM_005246386.4 7163 UTR 3 XP_005246443.1
XM_011510802.2 7163 UTR 3 XP_011509104.1
XM_011510803.2 7163 UTR 3 XP_011509105.1
XM_011510804.2 7163 UTR 3 XP_011509106.1
XM_011510805.2 7163 UTR 3 XP_011509107.1
XM_011510807.2 7163 UTR 3 XP_011509109.1
XM_011510808.2 7163 UTR 3 XP_011509110.1
XM_011510809.2 7163 UTR 3 XP_011509111.1
XM_011510810.2 7163 UTR 3 XP_011509112.1
XM_011510811.2 7163 UTR 3 XP_011509113.1
XM_011510812.2 7163 UTR 3 XP_011509114.1
XM_011510814.2 7163 UTR 3 XP_011509116.1
XM_011510815.2 7163 UTR 3 XP_011509117.1
XM_011510816.2 7163 UTR 3 XP_011509118.1
XM_011510817.2 7163 UTR 3 XP_011509119.1
XM_011510818.2 7163 UTR 3 XP_011509120.1
XM_011510819.2 7163 UTR 3 XP_011509121.1
XM_017003589.1 7163 UTR 3 XP_016859078.1
XM_017003590.1 7163 UTR 3 XP_016859079.1
XM_017003591.1 7163 UTR 3 XP_016859080.1
XM_017003592.1 7163 UTR 3 XP_016859081.1

View Full Product Details