Product Details

SNP ID
rs2231356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3118718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTCTTAGCGCATGCGCACTCC[C/T]GGCAGCTACGCGACCTGTGACCATG
Phenotype
MIM: 603156 MIM: 603453
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BPHL PubMed Links

Gene Details

Gene
BPHL
Gene Name
biphenyl hydrolase-like (serine hydrolase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302777.1 343 UTR 5 NP_001289706.1
NM_004332.3 343 UTR 5 NP_004323.2
Gene
RIPK1
Gene Name
receptor interacting serine/threonine kinase 1
There are no transcripts associated with this gene.

View Full Product Details