Product Details

SNP ID
rs2595650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:43944141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCACTCTGCTGCCTTGTGGAGAA[A/G]AGGCTGGGGTTCAAGTGATTCTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLR2J4 PubMed Links

Gene Details

Gene
POLR2J4
Gene Name
RNA polymerase II subunit J4, pseudogene
There are no transcripts associated with this gene.

Gene
UBE2D4
Gene Name
ubiquitin conjugating enzyme E2 D4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015983.3 Intron NP_057067.1
XM_006715734.3 Intron XP_006715797.1
XM_011515420.2 Intron XP_011513722.1
XM_011515422.2 Intron XP_011513724.1
XM_011515423.2 Intron XP_011513725.1
XM_017012292.1 Intron XP_016867781.1
XM_017012293.1 Intron XP_016867782.1
XM_017012294.1 Intron XP_016867783.1
XM_017012295.1 Intron XP_016867784.1

View Full Product Details