Product Details

SNP ID
rs2548776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65734742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGTGGAGTTGTAGTGACTGAA[A/G]TAATCACAGGTTGATGAAGCCTGCT
Phenotype
MIM: 611530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLN PubMed Links

Gene Details

Gene
NLN
Gene Name
neurolysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020726.4 Intron NP_065777.1
XM_005248559.2 Intron XP_005248616.1
XM_006714661.2 Intron XP_006714724.1
XM_017009673.1 Intron XP_016865162.1

View Full Product Details