Product Details

SNP ID
rs2656943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4878187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCGCAGCCACCCCCACAGCCAA[C/T]CCCTCCCTGGAGAACCGTGTCCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARRDC5 PubMed Links

Gene Details

Gene
ARRDC5
Gene Name
arrestin domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080523.1 Intron NP_001073992.1
XM_005259616.2 Intron XP_005259673.1
XM_011528181.2 Intron XP_011526483.1
XM_017027144.1 Intron XP_016882633.1
XM_017027145.1 Intron XP_016882634.1

View Full Product Details