Product Details

SNP ID
rs2917782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54329351 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAGTGCAAACAACACTTACTCTT[C/T]CATTGGTGAACATACAAAGCCACAA
Phenotype
MIM: 605252 MIM: 607051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLI PubMed Links

Gene Details

Gene
POLI
Gene Name
DNA polymerase iota
There are no transcripts associated with this gene.

Gene
STARD6
Gene Name
StAR related lipid transfer domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139171.1 627 Missense Mutation AAA,GAA K159E NP_631910.1
XM_011525821.1 627 Missense Mutation AAA,GAA K159E XP_011524123.1
XM_017025552.1 627 Missense Mutation AAA,GAA K100E XP_016881041.1
XM_017025553.1 627 Missense Mutation AAA,GAA K100E XP_016881042.1
XM_017025554.1 627 Missense Mutation AAA,GAA K77E XP_016881043.1

View Full Product Details