Product Details
- SNP ID
-
rs2232357
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:206866216 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACCTCCCACTAGTTCTTGGCAG[C/G]GAGTGGATGAGAAGTCTTGATATTG
- Phenotype
-
MIM: 605619
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL20
PubMed Links
Gene Details
- Gene
- IL20
- Gene Name
- interleukin 20
View Full Product Details