Product Details

SNP ID
rs2232657
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:26170873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATAAAGGAACTGGACAAAGAGA[A/G]CTTGCCTCCAACTCTAACTTCCAGG
Phenotype
MIM: 603272 MIM: 616698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNKSR1 PubMed Links

Gene Details

Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
There are no transcripts associated with this gene.

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
There are no transcripts associated with this gene.

Gene
ZNF593
Gene Name
zinc finger protein 593
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015871.4 649 UTR 3 NP_056955.2
XM_017001398.1 649 UTR 3 XP_016856887.1

View Full Product Details