Product Details

SNP ID
rs2232724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66552706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGAAAGTGCTGCTGCTGGGTC[G/T]GCAGACGCGATGGATAACGTGCAGC
Phenotype
MIM: 616074 MIM: 188250
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CKLF PubMed Links

Gene Details

Gene
CKLF
Gene Name
chemokine-like factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040138.2 144 UTR 5 NP_001035228.1
NM_016326.3 144 UTR 5 NP_057410.1
NM_016951.3 144 UTR 5 NP_058647.1
NM_181640.2 144 UTR 5 NP_857591.1
NM_181641.2 144 UTR 5 NP_857592.1
Gene
CKLF-CMTM1
Gene Name
CKLF-CMTM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202509.1 144 UTR 5 NP_001189438.1
NM_001204098.1 144 UTR 5 NP_001191027.1
NM_001204099.1 144 UTR 5 NP_001191028.1
Gene
TK2
Gene Name
thymidine kinase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details