Product Details

SNP ID
rs2233355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47580497 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTGGCTTTTGCCTTCTCCATTT[C/T]TTTTTTAAATATGCCTTTTCCTTCC
Phenotype
MIM: 614911 MIM: 603846 MIM: 609538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF4 PubMed Links

Gene Details

Gene
C1QTNF4
Gene Name
C1q and tumor necrosis factor related protein 4
There are no transcripts associated with this gene.

Gene
FAM180B
Gene Name
family with sequence similarity 180 member B
There are no transcripts associated with this gene.

Gene
KBTBD4
Gene Name
kelch repeat and BTB domain containing 4
There are no transcripts associated with this gene.

Gene
NDUFS3
Gene Name
NADH:ubiquinone oxidoreductase core subunit S3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004551.2 Intron NP_004542.1
Gene
PTPMT1
Gene Name
protein tyrosine phosphatase, mitochondrial 1
There are no transcripts associated with this gene.

View Full Product Details