Product Details
- SNP ID
-
rs2073200
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:42872089 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGAATCTCCCTGTGGCTCTAATGTA[A/T]TAACGTGGTTCCATTTATCTTTGGT
- Phenotype
-
MIM: 604960
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PACSIN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79192562] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PACSIN2
- Gene Name
- protein kinase C and casein kinase substrate in neurons 2
View Full Product Details