Product Details

SNP ID
rs2990233
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:155256326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCAGCGGCTGCATTGGCAGCTG[C/G]GGTTCGCACCGCAGGGTTGGAGAAG
Phenotype
MIM: 602989 MIM: 606913
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
There are no transcripts associated with this gene.

Gene
FAM189B
Gene Name
family with sequence similarity 189 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267608.1 1147 Intron NP_001254537.1
NM_006589.2 1147 Intron NP_006580.2
NM_198264.1 1147 Intron NP_937995.1
XM_005244845.1 1147 Intron XP_005244902.1
Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005698.3 1147 Missense Mutation CCA,GCA P331A NP_005689.2
NM_052837.2 1147 Missense Mutation CCA,GCA P305A NP_443069.1
XM_006711105.3 1147 Missense Mutation CCA,GCA P317A XP_006711168.1
XM_006711106.3 1147 Missense Mutation CCA,GCA P264A XP_006711169.1
XM_016999991.1 1147 UTR 3 XP_016855480.1

View Full Product Details